PYREX™ Filtering Flasks with Sidearm Tubulation
PYREX™ Filtering Flasks with Sidearm Tubulation
Actual product may vary
PYREX™ Filtering Flasks with Sidearm Tubulation
Tubulations for easy connections to filter pumps
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Closure Size
FIS10-180H4 L35.6 cm12
Catalog number FIS10-180H
Price (SGD)
-
Capacity (Metric):
4 L
Height (Metric):
35.6 cm
Closure Size:
12
Tubulations for easy connections to filter pumps
These flasks, with tubulation, are blown in special molds, in shapes designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers. Tubulation O.D. on sizes up to 1000 mL is approximately 10 mm. Tubulation O.D. for 2000 mL, and 4000 mL is approximately 12.5 mm.
Specifications
DescriptionStyle: Filtering; Top style: Heavy duty rim
Diameter (English) Outer7.79 in.
Diameter (Metric) Outer Tubing1.25 cm
Capacity (Metric)4 L
MaterialBorosilicate Glass
Graduation Range1500 to 3500 mL
Graduations500 mL
Height (English)14.01 in.
Height (Metric)35.6 cm
Neck StyleNarrow Mouth
No. per Case6/Cs.
No. per Pack1/Pk.
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Diameter (Metric) Outer198 mm
Closure Size12
Unit SizeEach