Digital Incubator
Actual product may vary
Digital Incubator
Digital incubator for laboratories that need a dependable, compact incubator for cultures, test kits, eggs and biologicals. Ideal for clinical, industrial and school laboratories, for physicians' offices, and as a general purpose warming cabinet.
Have Questions?
Catalog NumberDimensions (D x W x H) InteriorCapacity (English)
FIS10-180E10 x 12 x 10 in.0.7 cu. ft.
Catalog number FIS10-180E
Price (SGD)
627.00
Pack
Dimensions (D x W x H) Interior:
10 x 12 x 10 in.
Capacity (English):
0.7 cu. ft.
Price (SGD)
627.00
Pack
Temperature Range: Ambient +3°C minimum to a maximum of 93°C (200°F)
Digital Incubators (180 Series) all offer these standard features:
  • Digital PID microprocessor (Accuracy of ±0.5°C)
  • High grade aluminum interior.
  • Powder coated CR steel exterior.  
  • Resistive-tubular incoloy heating elements.
  • Solid insulated steel door
  • One fixed and one adjustable shelf
Specifications
Capacity (English)0.7 cu. ft.
AutoclavableNot Autoclavable
Capacity (Metric)19.8 L
Chamber MaterialAluminum
CO2 Sensor TechnologyNo Sensor
LightingNo Lighting
MaterialPowder-coated Cold Rolled Steel
Relative HumidityAmbient
Temperature Range (Metric)Ambient +3° to 93°C
TypeHatching Incubator
Temperature ControlBi-metal Thermostat
Temperature Stability±0.5°C
Dimensions (D x W x H) Interior10 x 12 x 10 in.
Width (English)13 in.
Width (English) Interior12 in.
Height (English)15 in.
Height (English) Interior10 in.
Depth (English)11 in.
Depth (English) Interior10 in.
Weight (English)30 lb.
Voltage115 V
Wattage270 W
Amperage2.1 A
Unit SizePack