PYREX™ Filtering Flasks with Sidearm Tubulation
Actual product may vary
PYREX™ Filtering Flasks with Sidearm Tubulation
Tubulations for easy connections to filter pumps
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Closure Size
FIS10-180D250 mL14.6 cm6
Catalog number FIS10-180D
Price (SGD)
448.00
Pack
Capacity (Metric):
250 mL
Height (Metric):
14.6 cm
Closure Size:
6
Price (SGD)
448.00
Pack
Tubulations for easy connections to filter pumps
These flasks, with tubulation, are blown in special molds, in shapes designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers. Tubulation O.D. on sizes up to 1000 mL is approximately 10 mm. Tubulation O.D. for 2000 mL, and 4000 mL is approximately 12.5 mm.
Specifications
DescriptionStyle: Filtering; Top style: Heavy duty rim
Diameter (English) Outer3.30 in.
Diameter (Metric) Outer Tubing1 cm
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Graduation Range75 to 250 mL
Graduations25 mL
Height (English)5.74 in.
Height (Metric)14.6 cm
Neck StyleNarrow Mouth
No. per Case18/Cs.
No. per Pack6/Pk.
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
Diameter (Metric) Outer84 mm
Closure Size6
Unit SizePack