Kimble™ KIMAX™ Culture Media Flasks
Kimble™ KIMAX™ Culture Media Flasks
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Culture Media Flasks
Screw-thread finish
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS10-093B250 mL
FIS10-093AA50 mL
FIS10-093A125 mL
Catalog number FIS10-093B
Price (SGD)
-
Capacity (Metric):
250 mL
Screw-thread finish
  • Includes long caps with cemented-in white rubber liners
Specifications
Diameter (English) Outer3.2 in.
Capacity (Metric)250 mL
MaterialBorosilicate Glass
For Use With (Application)Used to mix and store culture media, as well as with other culturing techniques
Graduation Range50 to 225 mL
Graduations25 mL
Height (English)5 in.
Height (Metric)12.8 cm
IncludesCap and white Rubber liner
Thread Size28-410
Closure TypeScrew
Diameter (Metric) Outer8.2 cm
Unit SizePK