PYREX™ Fernbach Flasks
Actual product may vary
PYREX™ Fernbach Flasks
Fernsbach culture flasks offer wide surface area is ideal for growth of organisms requiring a large surface area-to-volume ratio
Have Questions?
Catalog NumberProduct TypeNeck StyleClosure Type
FIS10-092Standard Fernbach FlaskBeadedNone
Catalog number FIS10-092
Price (SGD)
301.00
Each
Product Type:
Standard Fernbach Flask
Neck Style:
Beaded
Closure Type:
None
Price (SGD)
301.00
Each
  • Neck designs: plain, wide-mouth, beaded, or with screw cap
  • Capacity: 2800mL
  • Borosilicate glass
  • Nonsterile
Specifications
DescriptionStyle: Standard Fernbach; Top style: Beaded rim
Diameter (English) Outer8.05 in.
Capacity (Metric)2800 mL
MaterialBorosilicate Glass
For Use With (Application)For culturing organisms
Height (English)9.05 in.
Height (Metric)23 cm
Neck StyleBeaded
Product TypeStandard Fernbach Flask
Writing SurfaceYes
Mouth StyleWide
SterilityNon-sterile
Closure TypeNone
Diameter (Metric) Outer20.5 cm
Unit SizeEach