PYREX™ Short Neck Round Bottom Flask, Standard Taper Joint
PYREX™ Short Neck Round Bottom Flask, Standard Taper Joint
Actual product may vary
PYREX™ Short Neck Round Bottom Flask, Standard Taper Joint
Excellent chemical durability for contamination-free results
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (English)Height (Metric)Standard Taper Joint
FIS10-067DD250 mL4.7 in.12 cm19/22
FIS10-067CC100 mL3.7 in.9.5 cm19/22
Catalog number FIS10-067DD
Price (SGD)
173.00
CS
Add to cart
Capacity (Metric):
250 mL
Height (English):
4.7 in.
Height (Metric):
12 cm
Standard Taper Joint:
19/22
Price (SGD)
173.00
CS
Add to cart
With full length joints, but with short necks.
The 100, 250, and 500 mL flasks are replacement parts for organic chemistry kits (Cat. Nos. 6949 through 6949K).
Specifications
Diameter (English) Outer3.2 in.
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Height (English)4.7 in.
Height (Metric)12 cm
Neck StyleShort
No. per Case1/Cs.
No. per Pack1/Pk.
Standard Taper Joint19/22
Bottom ShapeRound
Diameter (Metric) Outer83 mm
Unit SizeCS