Check Order Status
Universal Precautions Lab Coats
Actual product may vary
Universal Precautions Lab Coats
Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure
Have Questions?
Catalog Number
Color
Size
Length (Metric)
FIS10-040H
White
Small
105.4 cm
Catalog number
FIS10-040H
Price (SGD)
150.00
PK
Add to cart
Color:
White
Size:
Small
Length (Metric):
105.4 cm
Price (SGD)
150.00
PK
Add to cart
For low to moderate exposure to fluids or chemicals
Traditional collar and knit cuffs
Six snaps
Three reinforced pockets
Back vent
Neck loop for hanging
Size/name label
Available in colors blue and white
Specifications
Size
Small
Material
SMS Fabric
Color
White
Cuff Style
Knit
Description
3-layer SMS fabric
Size (Metric) Chest
54.2 cm
Length (Metric)
105.4 cm