Eye Wash Bottle
Actual product may vary
Fisherbrand
Eye Wash Bottle
Eye wash bottle built-in eye cup provides immediate care.
Have Questions?
Catalog NumberCapacity (English)
FIS10-029-533.81 oz.
Catalog number FIS10-029-5
Price (SGD)
57.00
Each
Capacity (English):
33.81 oz.
Price (SGD)
57.00
Each
Eye wash bottle built-in eye cup provides immediate care.
  • Three stations available to suit all workstations: a single or a double station with 1000 mL (32 oz.) eyewash bottle(s) or a single station with a 500 mL (16 oz.) bottle
  • Shipped with empty bottles; can be filled with F24879-0032 Sterile Eye Wash Solution Refills
  • Instruction diagrams on bottle and station confirm how to use eyewash bottle
  • Includes multi-language directions on the station (English, French, German and Spanish)
Specifications
Capacity (English)33.81 oz.
Capacity (Metric)1000 mL
ColorNatural
Flow Rate1 gpm
For Use With (Application)Immediate Emergency Flushing of Eye Injuries
Height (English)11.62 in.
Height (Metric)29.51 cm
IncludesPlastic bottle, Built-in eye cup, captive snap-on cap, Multilanguage directions
MaterialLDPE
MountingFree Standing
Product TypeEyewash Bottle
Width (English)3.56 in.
Width (Metric)9.04 cm
Unit SizeEach