1x1 image pixel for data collection
Stainless Steel Wire Baskets
Stainless Steel Wire Baskets
Actual product may vary
Bel-Art
Stainless Steel Wire Baskets
Have Questions?
Catalog NumberHeight (Metric)
FIS10-028150 mm
Catalog number FIS10-028
Price (SGD)
154.00
EA
Add to cart
Height (Metric):
150 mm
Price (SGD)
154.00
EA
Add to cart
Specifications
Material18/10 E-Poli
Dimensions (L x W x H)180 x 180 x 150 mm
Length (Metric)180 mm
Width (Metric)180 mm
Height (Metric)150 mm
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link