Foam Swabs, Sterile
Actual product may vary
GE Healthcare
Foam Swabs, Sterile
For buccal and saliva samples.
Have Questions?
Catalog NumberQuantity
FIS09-923-375100 Pk.
Catalog number FIS09-923-375
Price (SGD)
418.00
Pack
Quantity:
100 Pk.
Price (SGD)
418.00
Pack
For buccal and saliva samples.
  • Sterile foam swabs for easy collection of buccal cells and saliva and transfer to e.g. QIAcard FTA card formats.
  • The nonabrasive head of the foam tipped applicator is the same size as the sample area on QIAcard FTA Cards to facilitate sample application.
Specifications
TypeFoam-tipped Applicator
For Use With (Application)For the collection of saliva and buccal cells
Quantity100 Pk.
Unit SizePack