QIAcard FTA Indicating Micro
Actual product may vary
GE Healthcare
QIAcard FTA Indicating Micro
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Micro with 1 sample area per card.
Have Questions?
Catalog NumberQuantity
FIS09-923-345100 Pk.
Catalog number FIS09-923-345
Price (SGD)
2,054.00
Pack
Quantity:
100 Pk.
Price (SGD)
2,054.00
Pack
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Micro with 1 sample area per card.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
DescriptionCollect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. With FTA, DNA stays in the matrix.
Isolation TechnologyFiltration
Sample TypeBuccal Cells, Saliva
For Use With (Application)Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples1
Product TypeIndicating FTA Micro Card
Quantity100 Pk.
Sample Volume125 μL
Unit SizePack