Glass Fiber Circles
Actual product may vary
Fisherbrand
Glass Fiber Circles
Faster flow, higher retention than cellulose filters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Grade
FIS09-804-42A4.2 cmG6
FIS09-804-55A5.5 cmG6
FIS09-804-42D4.2 cmG8
Catalog number FIS09-804-42A
Price (SGD)
28.00
Pack
Diameter (Metric):
4.2 cm
Grade:
G6
Price (SGD)
28.00
Pack
Faster flow, higher retention than cellulose filters
Used as prefilters and for gravimetric analyses. Suitable for many corrosive applications.
  • Low hygroscopicity
  • Chemically and biologically inert, with no organic binders
  • Offer higher retention efficiency of fine particles (<3μm in size)
  • Resist clogging for greater loading capacity than cellulose paper filters
  • Autoclavable and usable to 500°C (932°F)
  • Filters should be used flat: folding will break the glass fibers
Specifications
AutoclavableAutoclavable
MaterialGlass Fiber
Particle Retention1.6 μm
Flow RateFast
For Use With (Application)General purpose, air pollution monitoring, biochemical biological filtration cells, precipitated proteins, scintillation counting, radioimmunoassays
GradeG6
Temperature (English) Max.932°F
Temperature (Metric) Max500°C
Thickness (English)0.012 in.
Thickness (Metric)0.32 mm
Diameter (English)1.6 in.
Diameter (Metric)4.2 cm
Unit SizePack