Whatman™ Benchkote Surface Protector Sheets
Whatman™ Benchkote Surface Protector Sheets
Actual product may vary
Fisherbrand
Whatman™ Benchkote Surface Protector Sheets
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Have Questions?
Catalog NumberQuantity
FIS09-800-976100 Sheets
Catalog number FIS09-800-976
Price (SGD)
710.00
Pack
Add to cart
Quantity:
100 Sheets
Price (SGD)
710.00
Pack
Add to cart
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
  • Strong: Benchkote surface protector is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
  • Dimensions: 460 x 570mm
Specifications
Product TypeSurface Protector
MaterialPolyethylene
Length (Metric)460 mm
Width (Metric)570 mm
Dimensions (L x W)460 x 570mm
Backing MaterialPolyethylene
Thickness0.22mm
Quantity100 Sheets
Unit SizePack