Specifications
Filter TypeMicrofilter
FormatDisc
MaterialBorosilicate Glass
Particle Retention1.1 μm
Flow Rate3.6 cfm/ft2
For Use With (Application)Fine porosity and high flow for TSS, cell harvesting, and general laboratory applications
Grade161
Temperature (English) Max.932°F
Temperature (Metric) Max500°C
Thickness (English)0.012 in.
Thickness (Metric)0.32 mm
TypeGlass Microfiber Filter
Diameter (English)1.8 in.
Diameter (Metric)4.7 cm
Unit SizePack