Disposable Sterile Bottle-Top Filters, 0.45μm Membrane
Disposable Sterile Bottle-Top Filters, 0.45μm Membrane
Actual product may vary
Disposable Sterile Bottle-Top Filters, 0.45μm Membrane
Designed for use on glass or polystyrene media bottles with 33 and 45mm neck sizes. Corning™ Disposable Sterile Bottle-Top Filters are ideal for filtration of tissue culture media and components, biological fluids, other solutions.
Have Questions?
Catalog NumberVolume (Metric)Fits Bottle Neck Size (Metric)
FIS09-761-53500 mL45 mm
Catalog number FIS09-761-53
Price (SGD)
488.00
CS
Add to cart
Volume (Metric):
500 mL
Fits Bottle Neck Size (Metric):
45 mm
Price (SGD)
488.00
CS
Add to cart
  • Three sizes: 150 mL, 500 mL and 1,000 mL.
  • Low liquid retention funnel design.
  • Available in 33 mm and 45 mm neck sizes to fit most glass and plastic media storage bottles.
  • 45 mm neck sizes fit on Corning™ glass 1395 bottles, to be used on 2L and smaller sizes only.
  • Prefilters must be ordered separately.
  • Individually packaged.
  • Sterile.
  • Nonpyrogenic.
Specifications
ConnectorsHose
DisposableSingle-use
For Use With (Equipment)4.5 cm bottle neck
MaterialPolystyrene
Material (Membrane)Cellulose Acetate
Pore Size0.45 μm
SterilitySterile
Filtration Area33.2 cm2
Fits Bottle Neck Size (Metric)45 mm
For Use With (Application)Ideal for filtration of tissue culture media and components, biological fluids, other solutions
GraduatedNo
IncludesSquare membrane filter, graduated funnel of clear polystyrene with polyethylene neck orange adapter, and tubing adapter
No. per Case12/Cs.
No. per Pack1/Pk.
TypeBottle Top Vacuum Filter
Volume (Metric)500 mL
Diameter (Metric)7 cm
Unit SizeCS