Disposable Spinner Flask with Vent Cap and Angled Sidearms
Disposable Spinner Flask with Vent Cap and Angled Sidearms
Actual product may vary
Disposable Spinner Flask with Vent Cap and Angled Sidearms
PTFE Paddle Bushing was life tested for 2.5 months/24 hr./day at 125rpm
Have Questions?
Catalog NumberCapacity (Metric)
FIS09-761-31500 mL
Catalog number FIS09-761-31
Price (SGD)
7,733.00
Case
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
7,733.00
Case
Add to cart
  • The Corning disposable spinner flask system comes ready-to-use with a paddle and integrated magnet, reducing the need for time-consuming assembly, cleaning, or reassembly.
  • Molded from virgin polystyrene and sterile, each spinner flask system assures a clean sterile unit. No more concerns with detergent residues or contamination.
  • The paddle size and height are optimized for each vessel size. A unique integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers. Heat build-up in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface (125 mL and 500 mL flasks only).
Retrofit kits are available for converting older reusable spinner flasks to fit newer dual-bearing impellers.
Specifications
Capacity (English)0.132 gal.
Closure ColorOrange
Closure MaterialPolyethylene
Diameter (English) Outer3.4 in.
Diameter (Metric) Inner Side Arm3.8 cm
Diameter (Metric) Side Arm Cap5.4 cm
Capacity (Metric)500 mL
MaterialPolystyrene
Height (English)8 in.
Height (Metric)20.32 cm
IncludesTop cap and 2 angled side arms
No. per Case12/Cs.
No. per Pack1/Pk.
Side Arm Angle45°
Side Arm Openings2
Thread SizeGL 45
DisposableSingle-use
SterilitySterile
Closure TypeScrew cap, vented
Diameter (Metric) Outer87.3 mm
Unit SizeCase