Disposable Sterile Bottle-Top Filters with 0.22μm Membrane
Disposable Sterile Bottle-Top Filters with 0.22μm Membrane
Actual product may vary
Disposable Sterile Bottle-Top Filters with 0.22μm Membrane
Designed for use on glass or polystyrene media bottles with 33 and 45mm neck sizes. Corning™ Disposable Sterile Bottle-Top Filters are ideal for filtration of tissue culture media and components, biological fluids, other solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Filtration AreaFor Use With (Equipment)Material (Membrane)
FIS09-761-126150 mL13.6 cm24.5 cm bottle neckPES
FIS09-761-147500 mL33.2 cm24.5 cm bottle neckNylon
FIS09-761-112500 mL33.2 cm24.5 cm bottle neckPES
FIS09-761-1391000 mL54.5 cm24.5 cm bottle neckPES
Catalog number FIS09-761-126
Price (SGD)
1,340.00
PK
Add to cart
Volume (Metric):
150 mL
Filtration Area:
13.6 cm2
For Use With (Equipment):
4.5 cm bottle neck
Material (Membrane):
PES
Price (SGD)
1,340.00
PK
Add to cart
  • Three sizes: 150 mL, 500 mL and 1,000 mL.
  • Low liquid retention funnel design.
  • Available in 33 mm and 45 mm neck sizes to fit most glass and plastic media storage bottles.
  • 45 mm neck sizes fit on Corning™ glass 1395 bottles, to be used on 2L and smaller sizes only.
  • Prefilters must be ordered separately.
  • Individually packaged.
  • Sterile.
  • Nonpyrogenic.
Specifications
ConnectorsHose
DisposableSingle-use
For Use With (Equipment)4.5 cm bottle neck
MaterialPES
Material (Membrane)PES
Pore Size0.22 μm
SterilitySterile
Filtration Area13.6 cm2
For Use With (Application)Ideal for filtration of tissue culture media and components, biological fluids, other solutions
GraduatedYes
IncludesSquare membrane filter, graduated funnel of clear polystyrene with polyethylene neck yellow adapter, and tubing adapter
No. per Case48/Cs.
No. per Pack1/Pk.
TypeBottle Top Vacuum Filter
Volume (Metric)150 mL
Diameter (Metric)5 cm
Unit SizePK