Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Actual product may vary
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Thermo Scientific
Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeMaterial (Membrane)Diameter (Metric) MembraneVolume (Metric) Receiver
FIS09-741-2030.1 μmPES90 mm1000 mL
564-00200.2 μmPES50 mm50 mL
FIS09-741-880.2 μmPES50 mm50 mL
124-00450.45 μmPES50 mm115 mL
565-00100.1 μmPES50 mm150 mL
524-00200.2 μmPES50 mm115 mL
FIS09-741-000.2 μmPES50 mm115 mL
565-00200.2 μmPES50 mm150 mL
FIS09-741-010.2 μmPES50 mm150 mL
165-00450.45 μmPES50 mm150 mL
FKS100345500.45 μmPES50 mm150 mL
568-00100.1 μmPES75 mm250 mL
568-00200.2 μmPES50 mm250 mL
FIS09-741-040.2 μmPES50 mm250 mL
FKS104217910.2 μmPES50 mm250 mL
168-00450.45 μmPES50 mm250 mL
566-00100.1 μmPES75 mm500 mL
FIS09-741-2020.1 μmPES75 mm500 mL
566-00200.2 μmPES75 mm500 mL
FIS09-741-020.2 μmPES75 mm500 mL
FKS102290900.2 μmPES75 mm500 mL
569-00200.2 μmPES90 mm500 mL
FIS09-741-050.2 μmPES90 mm500 mL
166-00450.45 μmPES75 mm500 mL
FIS09-740-63E0.45 μmPES90 mm500 mL
567-00100.1 μmPES90 mm1000 mL
FKS126863660.1 μmPES90 mm1000 mL
567-00200.2 μmPES90 mm1000 mL
FIS09-741-030.2 μmPES90 mm1000 mL
FKS104117910.2 μmPES90 mm1000 mL
167-00450.45 μmPES90 mm1000 mL
FIS09-740-63C0.45 μmPES90 mm1000 mL
FKS102283400.45 μmPES90 mm1000 mL
121-00200.2 μmCellulose Nitrate50 mm115 mL
Catalog number FIS09-741-203
Price (SGD)
1,109.00
Case
Pore Size:
0.1 μm
Material (Membrane):
PES
Diameter (Metric) Membrane:
90 mm
Volume (Metric) Receiver:
1000 mL
Price (SGD)
1,109.00
Case
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).
  • Available in PES, CN, SFCA and Nylon membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
  • Available with Fit for Purpose Sterile documentation
  • Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see "Full Specifications" in the product table for item participation)
Specifications
ACT LabelYes
DisposableSingle-use
Material (Membrane)PES
Pore Size0.1 μm
SterilitySterile
No. per Case12
No. per Pack12
TypeFilter Unit
Volume (Metric) Receiver1000 mL
Volume (Metric) Upper1000 mL
Diameter (Metric) Membrane90 mm
Fit for PurposeSterile
Unit SizeCase