Syringe Filters - Sterile
Actual product may vary
Syringe Filters - Sterile
Low hold-up volume helps you to recover virtually all of your sample after filtering
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)Material (Membrane)
FIS09-720-0040.22 μm33 mmMCE
FIS09-720-30.22 μm13 mmPVDF
FIS09-719C0.2 μm25 mmNylon
FIS09-719D0.45 μm25 mmNylon
FIS09-720-0050.45 μm33 mmMCE
Catalog number FIS09-720-004
Price (SGD)
276.00
Pack
Pore Size:
0.22 μm
Diameter (Metric):
33 mm
Material (Membrane):
MCE
Price (SGD)
276.00
Pack
Low hold-up volume helps you to recover virtually all of your sample after filtering
Tested for 100% membrane and housing integrity, eliminating concerns about fluid lossIndividually wrapped
Hydrophilic PVDF Membrane
  • Hydrophilic membrane binds far less protein than nitrocellulose, nylon or PTFE
  • Ideal for filtration of protein-containing aqueous, mild organic solutions and other pharmaceutical/biological/biotech solutions
  • Sterilized by gamma radiation
Mixed Cellulose Ester (MCE) Membrane
  • Hydrophilic membrane has fair chemical and temperature resistance
  • EtOH sterilized
  • Use for clarifying or sterilizing aqueous solutions
Nylon Membrane
  • Excellent chemical resistance and low protein binding
  • Ideal for aqueous/biological application
  • Sterilized by gamma radiation
  • Hydrophilic
Specifications
Material (Membrane)MCE
Pore Size0.22 μm
Hold up Volume (After Air Purge)Low
TypeSyringe Filter
Diameter (Metric)33 mm
Unit SizePack