PYREX™ Soxhlet Extractor with Standard Taper Joints, Body Only
PYREX™ Soxhlet Extractor with Standard Taper Joints, Body Only
Actual product may vary
PYREX™ Soxhlet Extractor with Standard Taper Joints, Body Only
Bulb in siphon tube near the top of tube facilitates cycling of the siphoning action
Have Questions?
Catalog NumberCapacity (English)SizeStandard Taper Joint BottomStandard Taper Joint Top
FIS09-558B2.87 oz.Medium24/4045/50
Catalog number FIS09-558B
Price (SGD)
992.00
EA
Add to cart
Capacity (English):
2.87 oz.
Size:
Medium
Standard Taper Joint Bottom:
24/40
Standard Taper Joint Top:
45/50
Price (SGD)
992.00
EA
Add to cart
Bulb in siphon tube near the top of tube facilitates cycling of the siphoning action. The siphon tube is protected from accidental damage by the vapor tube. Standard taper joints at the top and bottom.
For glass extraction thimble with fritted disc only, see Cat. No. 33950.
Specifications
Product TypeExtraction Tube
For Use With (Equipment)Pyrex Soxhlet Apparatus Allihn Condenser
Capacity (Metric)85 mL
Capacity (English)2.87 oz.
SizeMedium
IncludesStandard taper joints, body only
For Use With (Application)Bulb in siphon tube near the top of tube facilitates cycling of the siphoning action
Standard Taper Joint Bottom24/40
Standard Taper Joint Top45/50
Diameter (English) Inner Chamber1.49 in.
Diameter (Metric) Inner Chamber3.8 cm
Height (English)11.92 in.
Height (Metric)30.3 cm
Length (English) Chamber5.11 in.
Length (Metric) Chamber13 cm
Unit SizeEA