PYREX™ Replacement Allihn Condenser
PYREX™ Replacement Allihn Condenser
Actual product may vary
PYREX™ Replacement Allihn Condenser
Replacement Allihn condenser with bulbs
Have Questions?
Catalog NumberStandard Taper Joint No
FIS09-557B45/50
Catalog number FIS09-557B
Price (SGD)
771.00
EA
Add to cart
Standard Taper Joint No:
45/50
Price (SGD)
771.00
EA
Add to cart
This is a PYREX replacement Allihn condenser with a inner Standard Taper joint on the bottom for the Cat. No. 3840 extractor system. The tubulation O.D. on the condenser is 10mm.
Specifications
Diameter (English) Outer Tubulation0.39 in.
Diameter (Metric) Outer Tubulation1 cm
For Use With (Equipment)3840-M (09-556B) 250 mL extractor system
Height (English)13 in.
Height (Metric)33.1 cm
No. of Bulbs4.0
Standard Taper Joint No45/50
MaterialBorosilicate Glass
TypeReplacement Allihn Condenser
Unit SizeEA