Drierite™ Gas Drying Unit with Hose Barbs
Actual product may vary
Drierite™ Gas Drying Unit with Hose Barbs
Drierite™ Gas Drying Unit with Hose Barbs
Drierite™ Gas Drying Unit with Hose Barbs
W.A. Hammond
Drierite™ Gas Drying Unit with Hose Barbs
Dries air and gases saturated with moisture to dew point. W A Hammond Drierite™Gas Drying Unit with Hose Barbs can be regenerated by removing from column and heating granules on tray in single layer
Have Questions?
Catalog NumberDiameter (Metric) Outer
FIS09-2046.65 cm
Catalog number FIS09-204
Price (SGD)
-
Diameter (Metric) Outer:
6.65 cm
Use anywhere to provide dry air or gas for applications such as: blanketing, sample preparation, instrument protection, reaction mixtures, purging, vacuum release, venting with dry air. Dries air and gases saturated with moisture to dew point. W A Hammond Drierite™ Gas Drying Unit with Hose Barbs can be regenerated by removing from column and heating granules on tray in single layer.
Color Change
The Indicating DRIERITE gives constant visual assurance of active desiccant. When active, Indicating DRIERITE is a distinct blue color. When exhausted, it turns pink. The zone between the two colors in the column may be a purple color and should be a sharp narrow band when the flow rate is at equilibrium. Regeneration will restore the drying capacity.
  • Air and gases saturated with moisture can be dried to dew point of approximately –73°C (–100°F)
  • Flow rates: 3.3L/m
  • Molded polycarbonate column with stainless-steel desicant supports and coil spring
  • Hose barbs for flexible tubing (1/4 in. - 3/8 in. I.D.)
  • Water capacity: 50g
  • Pressure drop: <0.01 psi at 200L/hr.
Specifications
Diameter (Metric) Outer6.65 cm
Product TypeLaboratory Gas Drying Unit
MaterialPolycarbonate column with stainless-steel desiccant supports and coil spring
For Use With (Application)Can be used anywhere to provide dry air or gas for applications such as blanketing, sample preparation, instrument protection, reaction mixtures, purging, vacuum release
Height (Metric)28.7 cm
Height (English)11.38 in.
Holds1.25 lb. 8-mesh Indicating Drietite
IncludesWith ∽567g of Drierite absorbent; hose barbs for flexible tubing (1.4 - 3/8 in. I.D.) (Cat. No. 09-204)
Diameter (English) Outer2.62 in.
Unit SizeEach