Chloride T2 Tablets
Actual product may vary
Lovibond
Chloride T2 Tablets
Lovibond Chloride T2 Tablets, Range: 0.5 to 25 mg/L, White, Composition: Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%, Format: Tablet, For Use With: Water analysis, MD600 colorimeter and the spectrodirect spectrophotometer
Have Questions?
Catalog NumberRange
FIS09-054-3110.5 to 25mg/L
Catalog number FIS09-054-311
Price (SGD)
-
Range:
0.5 to 25mg/L
Lovibond Chloride T2 Tablets, Range: 0.5 to 25mg/L, White, Composition: Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%, Format: Tablet, For Use With: Water analysis, MC500 colorimeter and SP600 spectrophotometer
Specifications
ColorWhite
CompositionPotassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%
pH Range4
FormatTablet
For Use With (Application)Water analysis
For Use With (Equipment)MC500 colorimeter and SP600 spectrophotometer
Range0.5 to 25mg/L
Shelf Life5 Years
Storage Requirements20°C
Test MethodSilver Nitrate/Turbidity
TypeChloride T2 Tablet
Unit SizePack