1x1 image pixel for data collection
Liston Bone Cutting Shears
Liston Bone Cutting Shears
Actual product may vary
Liston Bone Cutting Shears
For cutting bone and cartilage. Fisherbrand™ Bone Cutting Forceps are designed with heavy, strong, very sharp blades to cut through the toughest of bone.
Have Questions?
Catalog NumberLength (English)
FIS08-9905.5 in.
Catalog number FIS08-990
Price (SGD)
157.00
EA
Add to cart
Length (English):
5.5 in.
Price (SGD)
157.00
EA
Add to cart
Fisherbrand™ Bone Cutting Forceps are designed with heavy, strong, very sharp blades to cut through the toughest of bone. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage. Available in various blade lengths and sizes.
  • Strong, removable bar-spring makes shears self-opening
  • Stainless steel as per ASTM™ (F899-89) Standards
  • Blade length: 1.125 in. (30mm)
  • Sterilizable by EtO/gamma radiation
  • Rockwell Hardness: 50-55 HRC
Specifications
Blade StyleAngled
Length (English)5.5 in.
Length (Metric)13.97 cm
MaterialStainless Steel
AutoclavableAutoclavable
For Use With (Application)Dissecting
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link