Standard Dissecting Scissors
Actual product may vary
Standard Dissecting Scissors
Use these scissors for a variety of applications. Fisherbrand™ Standard Dissecting Scissors are forged from surgical-grade stainless steel. Available in multiple styles.
Have Questions?
Catalog NumberLength (English)Blade Style
FIS08-951-206 in.Straight
Catalog number FIS08-951-20
Price (SGD)
8.00
Each
Length (English):
6 in.
Blade Style:
Straight
Price (SGD)
8.00
Each
Use these scissors for a variety of applications. Fisherbrand™ Standard Dissecting Scissors are forged from surgical-grade stainless steel. Available in multiple styles.
  • Fisherbrand™ Standard Dissecting Scissors
  • Sharp blades for a clean and precise cut every time
  • English Pattern; Heavy-beveled edges for an aesthetic look
  • Electronically passivated
  • Autoclavable up to 135°
  • Surgical grade stainless steel
  • Can be sterilized by Eto/Gamma radiation
  • Rockwell Hardness (HRC): 46-50
  • Inspected per U.S
  • Government specifications for surgical instruments
  • Produced under ISO 9001, cGMP Guidelines
Specifications
Product TypeDissecting Scissors
Blade StyleStraight
GradeSurgical
Length (English)6 in.
Length (Metric)15.24 cm
MaterialStainless Steel
AutoclavableAutoclavable
For Use With (Application)Surgical, Precision
Tip StyleSharp/Sharp
Unit SizeEach