Dissecting Blunt-Pointed Forceps
Dissecting Blunt-Pointed Forceps
Actual product may vary
Dissecting Blunt-Pointed Forceps
Fisherbrand Blunt-Pointed Dissection Forceps are constructed from heavy duty stainless-steel, perfect for long lasting use.
Have Questions?
Catalog NumberLength (English)Length (Metric)MaterialTip Style
FIS08-8905 in.12.7 cmChrome-plated SteelStraight, serrated, blunt point
Catalog number FIS08-890
Price (SGD)
26.00
EA
Add to cart
Length (English):
5 in.
Length (Metric):
12.7 cm
Material:
Chrome-plated Steel
Tip Style:
Straight, serrated, blunt point
Price (SGD)
26.00
EA
Add to cart
Fisherbrand Blunt-Pointed Dissection Forceps are constructed from heavy duty stainless-steel, perfect for long lasting use.
  • Fisherbrand™ Medium-Pointed Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Tips grasp tissues and materials
  • Tips meet precisely when closed
  • Patterned-grip handles with guide pins (08-882 and 08-885)
  • Smooth handle without guide pins (08-900)
  • Passivated for corrosion resistance
  • ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Product TypeDissecting Forcep
AutoclavableAutoclavable
Certifications/ComplianceASTM
MaterialChrome-plated Steel
IncludesGuide pins
Latex FreeYes
Tip StyleStraight, serrated, blunt point
Length (English)5 in.
Length (Metric)12.7 cm
Unit SizeEA