Medium-Pointed Dissecting Forceps
Medium-Pointed Dissecting Forceps
Actual product may vary
Medium-Pointed Dissecting Forceps
Fisherbrand Medium-Pointed Dissection Forceps are used for grasping tissue and materials during dissection. ASTM™ standard stainless-steel
Have Questions?
Catalog NumberLength (English)Length (Metric)MaterialTip Style
FIS08-8824.5 in.11.4 cmStainless SteelCurved, serrated
Catalog number FIS08-882
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
4.5 in.
Length (Metric):
11.4 cm
Material:
Stainless Steel
Tip Style:
Curved, serrated
Fisherbrand Medium-Pointed Dissection Forceps are used for grasping tissue and materials during dissection. ASTM™ standard stainless-steel
  • Fisherbrand™ Medium-Pointed Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Tips grasp tissues and materials
  • Tips meet precisely when closed
  • Patterned-grip handles with guide pins (08-882 and 08-885)
  • Smooth handle without guide pins (08-900)
  • Passivated for corrosion resistance
  • ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Product TypeThoracic Forcep
AutoclavableAutoclavable
Certifications/ComplianceASTM
MaterialStainless Steel
FinishHigh Polish, Mirror Finish
For Use With (Application)Grasping of tissues and materials
Latex FreeYes
Tip StyleCurved, serrated
Length (English)4.5 in.
Length (Metric)11.4 cm
Unit SizeEA