Check Order Status
Glass Staining Dishes for 16 Slides
Actual product may vary
Glass Staining Dishes for 16 Slides
Staining jar holds 3 x 1 in. (75 x 25mm) slides vertically
Have Questions?
Catalog Number
Slide Count
FIS08-817
16
Catalog number
FIS08-817
Price (SGD)
60.00
EA
Add to cart
Slide Count:
16
Price (SGD)
60.00
EA
Add to cart
Staining jar holds 3 x 1 in. (75 x 25mm) slides vertically
Eight grooves, each wide enough to hold two slides back-to-back
With molded glass cover
Specifications
Closure Included
Closure Included
Closure Material
Glass
Bottom Shape
Flat
Material
Glass
Type
Coplin Staining Dish
Includes
Molded Glass Cover
Slide Count
16