Glass Staining Dishes for 10 Slides, with Ground Glass Cover
Glass Staining Dishes for 10 Slides, with Ground Glass Cover
Actual product may vary
Glass Staining Dishes for 10 Slides, with Ground Glass Cover
Hold up to 10 standard 3 x 1 in. (75 x 25mm) slides back-to-back
Have Questions?
Catalog NumberSlide Count
FIS08-81510
Catalog number FIS08-815
Price (SGD)
40.00
EA
Add to cart
Slide Count:
10
Price (SGD)
40.00
EA
Add to cart
Hold up to 10 standard 3 x 1 in. (75 x 25mm) slides back-to-back
  • Heavy molded glass Coplin staining dish holds slides vertically
  • Rectangular body with five grooves
Specifications
Closure IncludedClosure Included
Closure MaterialGlass
Bottom ShapeRound
MaterialGlass
TypeCoplin Staining Dish
Slide Count10
Height (Metric)11 cm
Height (English)4.31 in.
Unit SizeEA