Labs 3.5 - 5 kD Biotech CE Dialysis Kits
Actual product may vary
Labs 3.5 - 5 kD Biotech CE Dialysis Kits
Labs 3.5 - 5 kD Biotech CE Dialysis Kits
Spectrum
Labs 3.5 - 5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)
FIS08-801-24910 mm0.79 mL/cm vol./length
Catalog number FIS08-801-249
Price (SGD)
532.00
Each
Diameter (Metric):
10 mm
Length (Metric):
0.79 mL/cm vol./length
Price (SGD)
532.00
Each
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Product TypeDialysis Trial Kit
MWCO3.5 to 5 kDa
MaterialCellulose Ester
Diameter (Metric)10 mm
Diameter (English)0.39 in.
Length (Metric)0.79 mL/cm vol./length
Kit Contents1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
QuantityEach
For Use With (Application)Ideal for small, critical dialysis applications
Unit SizeEach