Spectra Mesh™ Woven, Nylon Filters
Actual product may vary
Spectrum
Spectra Mesh™ Woven, Nylon Filters
Consist of individual strands woven into a mesh screen
Have Questions?
Catalog NumberPore SizeDiameter (Metric)Thickness (Metric)
FIS08-801-1741 μm90 mm60 μm
Catalog number FIS08-801-17
Price (SGD)
740.00
Pack
Pore Size:
41 μm
Diameter (Metric):
90 mm
Thickness (Metric):
60 μm
Price (SGD)
740.00
Pack
Combines high tensile strength with flexibility
  • Water Affinity: Hydrophillic
  • Strength: High
  • Adsorption: High
  • Best Resistance: Corrosives
  • pH Resistance: pH 3-10
  • Thermal Stability: up to 180°C
  • Sterilization: Irradiation
Specifications
AdsorptionHigh
Certifications/ComplianceISO 9001:2008
MaterialNylon
Pore Size41 μm
SterilitySterile
HydrophobicityHydrophilic
Open Area33%
pH Range3 to 10
Size (Metric) Opening41μm
Temperature Stability180°C
Thickness (Metric)60 μm
TypeSpectra Mesh Woven Filter
Diameter (English) Inside3.54 in.
Diameter (Metric)90 mm
Diameter (Metric) Inside9 cm
Unit SizePack