Chambered  Cell Culture Slides
Actual product may vary
Chambered  Cell Culture Slides
Chambered  Cell Culture Slides
Chambered Cell Culture Slides
Packaged in disposable incubation trays. Falcon™ Chambered Cell Culture Slides are ideal for in situ DNA amplification/hybridization, tissue and tumor typing/pathology, or any study requiring a small, accessible culture area.
Have Questions?
Catalog NumberCulture AreaWellsNo. per CaseVolume (Metric) Working
FIS08-774-260.7 cm28240.7 to 0.8 mL
Catalog number FIS08-774-26
Price (SGD)
1,075.00
Case
Culture Area:
0.7 cm2
Wells:
8
No. per Case:
24
Volume (Metric) Working:
0.7 to 0.8 mL
Price (SGD)
1,075.00
Case
  • Simplify analysis: culture, fix, stain and  examine your samples under a microscope without cell transfer
  • Polystyrene chamber affixed to a specially cleaned and triple-rinsed glass microscope slide (25 × 75mm)
  • Safety Removal Tool separates wells from slide; gasket remains with plastic vessel, prevents cross-contamination and doesn't interfere with further processing
  • Sterile
  • Culture up to 8 different conditions on a single glass microscope slide
  • Numeric labeling; 4-well, and 8-well formats available
Specifications
TypeCell Culture Slide
Wells8
Culture Area0.7 cm2
Volume (Metric) Working0.7 to 0.8 mL
For Use With (Application)In situ DNA amplification or hybridization, tissue and tumor typing or pathology, or any study requiring a small, accessible culture area
IncludesHandled polystyrene lid, safety removal tool
No. per Case24