Cell Strainers
Cell Strainers
Actual product may vary
Cell Strainers
Rapidly isolates primary cells to consistently obtain a uniform single-cell suspension from tissues. Falcon™ Cell Strainers are easy to use and ideal for use with stem cells and primary cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMesh SizeColorSterility
FIS08-771-140 μmBlueSterile
FIS08-771-19100 μmYellowSterile
FIS08-771-270 μmWhiteSterile
Catalog number FIS08-771-1
Price (SGD)
504.00
CS
Add to cart
Mesh Size:
40 μm
Color:
Blue
Sterility:
Sterile
Price (SGD)
504.00
CS
Add to cart
  • A faster and easier alternative to gauze filtration in procedures involving dissociation of cells from either clumps or primary tissues
  • Consistently obtain a more uniform single-cell suspension
  • Three nylon mesh pore sizes for optimal performance in a variety of applications
  • Sterilized and conveniently accessible in individual packaging
  • Extended lip on strainer enables aseptic handling with forceps
  • Strainers are made of a strong nylon mesh with 40, 70, or 100 μm pores that are evenly spaced for consistent results
  • Molded color-coded polypropylene frame with tab enables easy handling
  • Features
    Sterile
  • Individually packaged
Specifications
Mesh MaterialNylon
DisposableSingle-use
ColorBlue
For Use With (Application)Designed for rapidly isolating primary cells to consistently obtain a uniform single cell suspension from tissues
IncludesExtended lip on strainer, polypropylene frame with tab
MaterialPolypropylene
Product TypeCell Strainer
SterilitySterile
For Use With (Equipment)Conical Tubes, 50 mL Falcon
Mesh Size40 μm
Unit SizeCS