Adjustable Microplate Tilting Stand
Actual product may vary
Adjustable Microplate Tilting Stand
Adjustable Microplate Tilting Stand
Fisherbrand
Adjustable Microplate Tilting Stand
Adjusts to most convenient angle for precise additions of liquid components to microplate wells. Bel-Art™ Adjustable Microplate Tilting Stand allows you to easily judge which wells have received liquid addition.
Have Questions?
Catalog NumberHolds
FIS08-761Microplates
Catalog number FIS08-761
Price (SGD)
459.00
Each
Holds:
Microplates
Price (SGD)
459.00
Each
  • Accurate, repetitive aliquoting in microplate wells is simple by slanting plates to an optimal angle by placing the plate on the adjustable mounting stand
  • Adding aliquots is simple with the microplate slanted at an angle that is optimal to the user's working habit and convenience
  • The slanted plate permits the user to track which wells have received aliquots by visibly contrasting the menisci between adjoining wells
  • An epoxy coated steel base provides stability, and the hand adjustable clamping screws maintain the desired angle
  • Holds all sizes of microplates
  • Max tilt 45°
  • Pivots on 2 screws
  • L x W x H: 11.4 x 16.5 x 5.7cm (4.5 x 6.5 x 2.25 in.)
Specifications
AutoclavableNot Autoclavable
ColorGray, Clear/White
HoldsMicroplates
MaterialEpoxy-coated Steel, Acrylic
Positions1
ShapeRectangular
StackableNo
For Use With (Equipment)All sizes of microplates
Size114 x 165 x 57 mm
Dimensions (L x W x H)6.4 x 4.4 x 2.2 in. (16.5 x 11.4 x 5.7cm)
Length (Metric)16.5 cm
Length (English)6.4 in.
Width (English)4.4 in.
Height (English)2.2 in.
Product TypeMicroplate Rack
Height (Metric)5.7 cm
Array1 x 1
Width (Metric)11.4 cm
Unit SizeEach