PYREX™ Evaporating Dishes
PYREX™ Evaporating Dishes
Actual product may vary
PYREX™ Evaporating Dishes
Withstand temperatures to 230°C (446°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (English)Height (English)
FIS08-710C290 mL4.13 in.2.16 in.
FIS08-710A140 mL4.13 in.1.77 in.
FIS08-710B170 mL3.54 in.1.96 in.
Catalog number FIS08-710C
Price (SGD)
1,214.00
CS
Add to cart
Capacity (Metric):
290 mL
Diameter (English):
4.13 in.
Height (English):
2.16 in.
Price (SGD)
1,214.00
CS
Add to cart
Withstand temperatures to 230°C (446°F)
These dishes have rugged beaded rims and are of standard medium depth.
Specifications
Bottom ShapeFlat
Capacity (Metric)290 mL
MaterialBorosilicate Glass
ShapeRound
TypeEvaporating Dish
No. per Case6/Cs.
No. per Pack1/Pk.
Temperature Range (English)446°F
Temperature Range (Metric)230°C
Height (Metric)55 mm
Height (English)2.16 in.
Diameter (English)4.13 in.
Diameter (Metric)105 mm
Unit SizeCS