Check Order Status
Labs Spectra/Por™ 6 1000 D MWCO Standard RC Pre-wetted Dialysis Kits
Fisherbrand
Labs Spectra/Por™ 6 1000 D MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
Have Questions?
Catalog Number
Diameter (Metric)
Length (Metric)
FIS08-700-196
24 mm
4.6 mL/cm vol./length
Catalog number
FIS08-700-196
Price (SGD)
0.00
EA
Add to cart
Diameter (Metric):
24 mm
Length (Metric):
4.6 mL/cm vol./length
Price (SGD)
0.00
EA
Add to cart
Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
Membrane packaged wet in preservative solution of 0.05% sodium azide
Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
Include a pair of closures and convenient tubing opening picks
Specifications
MWCO
1 kDa
Diameter (Metric)
24 mm
Length (Metric)
4.6 mL/cm vol./length
Quantity
1 m Tubing/Kit