Imetra Diamond Scribes
Actual product may vary
Imetra Diamond Scribes
Diamond tip writing instrument provides comfortable grip for etching glass and other surfaces
Have Questions?
Catalog NumberQuantity
FIS08-6751 Ea.
Catalog number FIS08-675
Price (SGD)
-
Quantity:
1 Ea.
Diamond tip writing instrument provides comfortable grip for etching glass and other surfaces
Sturdy etching instrument designed to look and feel like a pen
  • Diamond tip
  • Aluminum shaft
  • Clip
  • Barrel's shape: Hexagonal
  • Cap included
Specifications
Product TypeScribe Pen
ColorSilver
Tip MaterialDiamond
For Use With (Application)Glass, ceramic, metal, and other.
Quantity1 Ea.
Unit SizeEach