▸
Protein Analysis Reagents
▸
Dialysis and Desalting
▸
Dialysis Tubing
▸
Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
Actual product may vary
Spectrum
Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
Selected MWCOs from 100 to 100,000 Daltons
Have Questions?
Change view
Catalog Number
Diameter (Metric)
MWCO
FIS08-671-41
20 mm
100 kDa
FIS08-671-25
20 mm
0.5 kDa
FIS08-671-27
20 mm
1 kDa
FIS08-671-31
20 mm
5 kDa
FIS08-671-39
10 mm
100 kDa
FIS08-670-264
20 mm
25 kDa
6 Options
Catalog number
FIS08-671-41
Price (SGD)
1,788.00
Each
Add to cart
Diameter (Metric):
20 mm
MWCO:
100 kDa
Price (SGD)
1,788.00
Each
Add to cart
Selected MWCOs from 100 to 100,000 Daltons
CE tubing membranes are ideal for desalting, purification, and molecular separation
Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution
Specifications
Product Type
Spectra/Por Dialysis Membrane Tubing
MWCO
100 kDa
Material
Cellulose Ester
Diameter (Metric)
20 mm
Diameter (English)
0.78 in.
Length (English)
33 ft.
Length (Metric)
10 m
Formulation
Supplied in 0.1% sodium azide solution
Temperature Range (English)
39.2°F to 98.6°F
Temperature Range (Metric)
4°C to 37°C
Quantity
Each
For Use With (Application)
Ideal for desalting, purification, and molecular separation