Spectra/Mesh Woven Polymer Macrofiltration Squares: N (Nylon)
Actual product may vary
Spectrum
Spectra/Mesh Woven Polymer Macrofiltration Squares: N (Nylon)
Combines high tensile strength with flexibility
Have Questions?
Catalog NumberOpen AreaSize (Metric) Opening
FIS08-670-19936%70μm
Catalog number FIS08-670-199
Price (SGD)
1,251.00
Pack
Open Area:
36%
Size (Metric) Opening:
70μm
Price (SGD)
1,251.00
Pack
Combines high tensile strength with flexibility
Ideal for straining, filtering and screening solutions or suspensions containing high-molecular-weight molecules or particles.
  • Nonabsorbent
  • Combines high tensile strength with flexibility
Specifications
AdsorptionHigh
Certifications/ComplianceISO 9001:2008
MaterialNylon
SterilitySterile
HydrophobicityHydrophilic
Open Area36%
pH Range3 to 10
Size (Metric) Opening70μm
Temperature Stability180°C
Thickness (Metric)70 μm
TypeSpectra Mesh Woven Filter
Diameter (English)4.65 in.2
Unit SizePack