Dricycler
Actual product may vary
Dricycler
Dricycler
Dricycler
Boekel Scientific
Dricycler
The Dricycler with automatic desiccant regenerates the silica gel desiccant every five hours. Ideal for storing crucibles, storing reagents, and drying specimens.
Have Questions?
Catalog NumberSize
FIS08-645-32Small
Catalog number FIS08-645-32
Price (SGD)
9,977.00
Each
Size:
Small
Price (SGD)
9,977.00
Each
  • Automatically regenerates desiccant every five hours
  • Adjustable shelves
  • Supplied with 2 shelves
  • Hand crafted from stainless steel and glass
Specifications
Cabinet StyleFront-loading
MaterialStainless Steel, Glass
Product TypeDricycler
For Use With (Application)Storing reagents or soil samples, drying biological specimens, storage sludge samples, cooling down from 40° to 150°C.
IncludesAdjustable shelves, neoprene door gasket, full-length door hinges
No. of Shelves2
Shelf Positions1/2 in. Increments
SizeSmall
Warranty2 Years
Dimensions (L x W x H)12.5 x 16 x 12.25 in. (31.4 x 40.6 x 31 cm)
Length (English)12.5 in.
Length (English) Chamber10 in.
Length (Metric)31.4 cm
Length (Metric) Chamber25.4 cm
Width (English)16 in.
Width (English) Chamber11.5 in.
Width (Metric)40.6 cm
Width (Metric) Chamber29.2 cm
Height (English)12.25 in.
Height (English) Chamber12 in.
Height (Metric)31 cm
Height (Metric) Chamber30.5 cm
Weight (English)26 lb.
Weight (Metric)11.8 kg
Voltage115 VAC
Wattage35 W