Fiberglass Desiccator Cabinet
Fiberglass Desiccator Cabinet
Actual product may vary
Labconco
Fiberglass Desiccator Cabinet
Withstands 29 in. Hg (0.1MPa) pressure
Have Questions?
Catalog NumberNo. of Shelves
FIS08-6442
Catalog number FIS08-644
Price (SGD)
-
No. of Shelves:
2
  • Vacuum release needle valve equalizes chamber pressure gradually
  • Made of molded polyethylene
  • Clear tempered glass door
  • Nine molded shelf supports permit flexible shelf arrangements
  • Can be stacked on top of each other using accessory Stacking Rack 16-307-242
Specifications
ColorClear
MaterialPolyethylene
For Use With (Application)Provide low humidity environment for the storage of moisture-sensitive materials including thin layer chromatography plates, halide salt cells for IR spectrophotometers and chemical standards
IncludesTwo 12 x 12 in. shelves with twelve 0.875 in. holes in each, clear tempered glass door, needle valve, desiccant pan and wire support stand
No. of Shelves2
Width (English)17 in.
Width (Metric)43 cm
Height (English)15.38 in.
Height (Metric)39 cm
Depth (English)13.5 in.
Depth (Metric)34 cm
Unit SizeEach