Acrylic Desiccator Cabinets
Acrylic Desiccator Cabinets
Actual product may vary
Acrylic Desiccator Cabinets
Dust- and moisture-free storage in refrigerators, freezers and on benchtops
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)No. of Shelves
FIS08-642-23C45.7 cm4 Removable Adjustable and Vented
FKS1199786417.8 cm2 Removable Adjustable and Vented
FIS08-642-23B30.5 cm3 Removable Adjustable and Vented
FKS1190787430.5 cm3 Removable Adjustable and Vented
FKS1191787445.7 cm4 Removable Adjustable and Vented
Catalog number FIS08-642-23C
Price (SGD)
2,104.00
Each
Add to cart
Height (Metric):
45.7 cm
No. of Shelves:
4 Removable Adjustable and Vented
Price (SGD)
2,104.00
Each
Add to cart
Dust- and moisture-free storage in refrigerators, freezers and on benchtops
  • Ideal for storing anhydrous and hygroscopic biological and chemical compounds, as well as electronic assemblies and any components requiring controlled storage conditions
  • Neoprene gasket on door ensures tight seal
  • Door is secured with two stainless-steel latches (large size has three latches) and full-height hinge
  • Removable, lipped tray holds solid desiccant
  • Shelves are removable, adjustable and vented to maximize airflow
Specifications
AutoclavableNot Autoclavable
ColorClear
Cabinet StyleFront-loading
MaterialAcrylic
For Use With (Application)Ideal for storing anhydrous and hygroscopic biological and chemical compounds, as well as electronic assemblies and any components requiring controlled storage conditions
IncludesNeoprene gasket, two stainless steel latches (large size has three latches) and full-height hinge, removable solid desiccant lipped tray, vented shelves
No. of Shelves4 Removable Adjustable and Vented
Dimensions (L x W)12 x 12 in. (30.4 x 30.4 cm)
Length (English)12 in.
Length (Metric)30.5 cm
Width (English)12 in.
Width (Metric)30.5 cm
Height (English)18 in.
Height (Metric)45.7 cm
Unit SizeEach