Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Actual product may vary
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Thermo Scientific
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
View contents easily with these autoclavable polypropylene desiccators, featuring a transparent PC cover and robust body resistant to all solid desiccants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Diameter (Metric) Inner
FIS08-642262 mm246 mm
FIS08-642-15149 mm149 mm
FKS10548842149 mm149 mm
Catalog number FIS08-642
Price (SGD)
491.00
Each
Height (Metric):
262 mm
Diameter (Metric) Inner:
246 mm
Price (SGD)
491.00
Each
View contents easily with Thermo Scientific™ Autoclavable Polypropylene Desiccators, featuring a robust body and high resistance to all solid desiccants. These desiccators have a transparent polycarbonate cover. The small size (5315-0150) comes with a 140 mm aluminum plate. The plate for the large size (5309-0250) is sold separately.
  • Clear polycarbonate (PC) cover has recessed rim to keep grease off benchtop when cover is set down
  • No gasket required, only grease is needed
  • Large desiccator (Cat. No. 5310-0250) takes any standard 230mm desiccator plate such as Cat. No. 5312-0230 (sold separately)
  • Autoclavable; repeated autoclaving will reduce the life of the product
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
AutoclavableAutoclavable
DescriptionDesiccator PP/PC 250mm
Cabinet StyleTop Loading
MaterialPolypropylene, Polycarbonate
Max. Clearance Above Plate5.6 in.
No. of Shelves1 (sold separately)
No. per Pack1
Height (English)10.32 in.
Height (Metric)262 mm
Diameter (English) Inner9.68 in.
Diameter (English) Outer13 in.
Diameter (Metric) Inner246 mm
Diameter (Metric) Outer330 mm
Unit SizeEach