Porcelain Desiccator Plates
Porcelain Desiccator Plates
Actual product may vary
Porcelain Desiccator Plates
One large center hole
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) OuterDiameter (Metric) OuterProduct Type
FIS08-641C9.05 in.23 cmDesiccator Plate with 5 mm Perforations
FIS08-641A5.51 in.14 cmDesiccator Plate with 5 mm Perforations
Catalog number FIS08-641C
Price (SGD)
-
Diameter (English) Outer:
9.05 in.
Diameter (Metric) Outer:
23 cm
Product Type:
Desiccator Plate with 5 mm Perforations
  • Without feet
  • Glazed on one side
Specifications
Diameter (Metric) Outer23 cm
Product TypeDesiccator Plate with 5 mm Perforations
MaterialPorcelain
For Use With (Equipment)Desiccator
Diameter (English) Outer9.05 in.
Unit SizeEA