Desi-Can™ Reusable Desiccant Canister
Desi-Can™ Reusable Desiccant Canister
Actual product may vary
Desi-Can™ Reusable Desiccant Canister
Desi-Can™ Reusable Desiccant Canister
Desi-Can™ Reusable Desiccant Canister
Bel-Art
Desi-Can™ Reusable Desiccant Canister
For use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case.
Have Questions?
Catalog number FIS08-594-19
Price (SGD)
89.00
EA
Add to cart
Price (SGD)
89.00
EA
Add to cart
For use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case
  • One canister of Reusable Desi-Can Canister maintains a safe level of humidity in 0.09 cubic meter (3 cubic ft.) of space
  • Can be regenerated in 121°C (250°F) oven in about 3 hours for unlimited life
  • Monitor window shows blue when recharged and pink when saturated
Specifications
Diameter (Metric) Outer10.1 cm
Product TypeDesiccator Canister
MaterialSilica Gel
Height (Metric)1.27 cm
Height (English)0.5 in.
Unit SizeEA