Nalgene™ Fluorocarbon (PFA) Resin Plastic Graduated Cylinder
Actual product may vary
Nalgene™ Fluorocarbon (PFA) Resin Plastic Graduated Cylinder
Nalgene™ Fluorocarbon (PFA) Resin Plastic Graduated Cylinder
Nalgene™ Fluorocarbon (PFA) Resin Plastic Graduated Cylinder
Thermo Scientific
Nalgene™ Fluorocarbon (PFA) Resin Plastic Graduated Cylinder
Measure aggressive chemicals and other liquids without the risk of breaking with the extraordinary chemical resistance of these graduated cylinders.
Have Questions?
Catalog NumberCapacity (Metric)
FIS08-572-8D100 mL
Catalog number FIS08-572-8D
Price (SGD)
2,238.00
Case
Capacity (Metric):
100 mL
Price (SGD)
2,238.00
Case
Measure aggressive chemicals and other liquids without the risk of breaking with the Thermo Scientific™ Nalgene™ Fluorocarbon (PFA) Resin Graduated Cylinder, featuring extraordinary chemical resistance. This cylinder tolerates a wide temperature range - from -270°C to +250°C (-454°F to 482°F). Graduations at 2mL increments with numbered graduations every 10mL are molded-in and won't rub off.
  • Low in extractables—won't contaminate contents
  • Generous pour spout provides ease in dispensing liquids
  • Autoclavable/Graduated/PFA
Specifications
Capacity (Metric)100 mL
MaterialPFA
Tolerance±1.5 mL
Diameter (English)1.55 in.
Scale8mL to 100mL in 2mL increments
Diameter (Metric)39.4 mm
Quantity1
Graduations2 mL
AutoclavableAutoclavable
Limit of Error±1.50mL
ShapeRound
Max. Temperature (English)482°F
Base TypeRound
Height (English)6.9 in.
Height (Metric)175.3 mm
Product TypeGraduated Cylinder
Max. Temperature (Metric)250°C
Diameter (English) Inner1.2 in.
Diameter (Metric) Inner30.5 mm
Unit SizeCase