Serialized Class A Cylinders with Certificate of Calibration
Actual product may vary
Serialized Class A Cylinders with Certificate of Calibration
Calibrated To Deliver.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS08-557DC100 mL1 mL
FIS08-557BC25 mL0.2 mL
FIS08-557EC250 mL2 mL
FIS08-557FC500 mL5 mL
FIS09-730-311000 mL10 mL
Catalog number FIS08-557DC
Price (SGD)
313.00
Each
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (SGD)
313.00
Each
Calibrated To Deliver.
  • Traceability
  • White scale
Specifications
Base TypeHexagonal
CalibrationTo Deliver
Scale ColorWhite
ShapeRound
ClassA
Height (English)10 in.
Height (Metric)255 mm
Tolerance Metric±0.4 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)100 mL
IncludesIndividual Fused-In Serial Numbers, Traceable NIST Certificate
Graduations1 mL
Subdivision1mL
Unit SizeEach