Class B Cylinders with Double Metric Scales
Class B Cylinders with Double Metric Scales
Actual product may vary
Class B Cylinders with Double Metric Scales
Calibrated To Deliver
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsTolerance MetricSubdivision
FIS08-555G1000 mL10 mL±6 mL10mL
FIS08-555A10 mL0.2 mL±0.2 mL0.2mL
FIS08-555B25 mL0.5 mL±0.34 mL0.5mL
FIS08-555D100 mL1 mL±1 mL1mL
FIS08-555C50 mL1 mL±0.5 mL1mL
FIS08-555E250 mL2 mL±2 mL2mL
Catalog number FIS08-555G
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
10 mL
Tolerance Metric:
±6 mL
Subdivision:
10mL
Calibrated To Deliver
  • Two sets of white ceramic graduations for reading up or down
  • Heavy base
  • Pouring lip
  • Accuracy assured even at lowest graduations; first 10% of volume at bottom is not graduated
Specifications
Base TypeRound
CalibrationTo Deliver
Scale ColorWhite
ShapeRound
ClassB
Tolerance Metric±6 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)1000 mL
For Use With (Application)Accuracy assured even at lowest graduations
ScaleDual
Graduations10 mL
AutoclavableAutoclavable
Limit of Error+/- 6.0mL
Subdivision10mL
Unit SizeEA