1x1 image pixel for data collection
PYREX™ Certified/Serialized Class A Cylinders with Metric Scale, TD
PYREX™ Certified/Serialized Class A Cylinders with Metric Scale, TD
Actual product may vary
PYREX™ Certified/Serialized Class A Cylinders with Metric Scale, TD
Calibrated To Deliver
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS08-553E25 mL0.2 mL
FIS08-553B100 mL1 mL
FIS08-553A50 mL1 mL
Catalog number FIS08-553E
Price (SGD)
471.00
CS
Add to cart
Capacity (Metric):
25 mL
Graduations:
0.2 mL
Price (SGD)
471.00
CS
Add to cart
  • Calibrated “To Deliver” in accordance with ASTM E-1272 and ASTM E-542.
  • Each cylinder is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards.
  • Permanent white graduations are easy to read.
  • Bumper guards are supplied with 25 mL through 2000 mL inclusive.
  • The 10 mL size has a funnel top.
  • Tested and calibrated in an ISO/IEC 17025 certified laboratory.
Specifications
Base TypeHexagonal
CalibrationTo Deliver
Scale ColorWhite
ShapeRound
SterilityNon-sterile
Certifications/ComplianceASTM E1272, ASTM E542, ASTM E694
ClassSerialized/Certified Class A
Height (English)7.55 in.
Height (Metric)194 mm
Tolerance Metric±0.17 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Diameter (English) Outside0.7 in.
Diameter (Metric) Outside18 mm
Capacity (Metric)25 mL
IncludesCase of 1
ScaleSingle
Graduations0.2 mL
AutoclavableAutoclavable
Subdivision0.2 mL
Unit SizeCS
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link