Search Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FIS08-549-12A | 25 mL |
Designed for microchemical work. The heavy duty, wide mouth facilitates filling and mixing. Easy-to read graduation line is sharp and permanent. Supplied with interchangeable standard taper ground glass stoppers.